Learn More
Cayman Chemical β-Phellandrene, Purity: ≥95%, CAS: 555-10-2, 100 mg
SDP
Catalog No. 502947864
Encompass_Preferred
A cyclic monoterpene with apoptotic activity; cytotoxic to RAW 264.7 cells at 50-200 µg/ml; induces apoptosis in RAW 264.7 cells
Catalog No. 50-294-7864 Supplier Cayman Chemical Supplier No. 33905100 MG
A cyclic monoterpene with apoptotic activity; cytotoxic to RAW 264.7 cells at 50-200 µg/ml; induces apoptosis in RAW 264.7 cells