Learn More
Cayman Chemical Phellandrene 50mg
SDP
Catalog No. 502947097
Encompass_Preferred
A cyclic monoterpene with apoptotic activity; cytotoxic to RAW 264.7 cells at 50-200 UG/ml; induces apoptosis in RAW 264.7 cells
Catalog No. 50-294-7097 Supplier Cayman Chemical Supplier No. 3390550 MG
A cyclic monoterpene with apoptotic activity; cytotoxic to RAW 264.7 cells at 50-200 UG/ml; induces apoptosis in RAW 264.7 cells