Learn More
Medchemexpress LLC HY-N2110 5mg Medchemexpress, Phellopterin CAS:2543-94-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN21105MG
Encompass_Preferred
Medchemexpress, HY-N2110 5mg Phellopterin CAS:2543-94-4 Phellopterin is a natural product isolated from P. trifoliata. Phellopterin reduces TNF-alpha-induced VCAM-1 expression through regulation of the Akt and PKC pathway, which contributes to inhibit the adhesion of monocytes to endothelium. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2864