Learn More
Cayman Chemical Phellodendrine (chloride), Purity: ≥98%, CAS: 104112-82-5, 10 mg
SDP
Catalog No. 502911302
Encompass_Preferred
An alkaloid; inhibits AChE activity (IC50 = 36.51 µM and scavenges ABTS radicals in cell-free assays from 12.5-100 µM; induces apoptosis in PANC-1 pancreatic cancer cells and reduces tumor growth in a PANC-1 mouse xenograft model; inhibits the GVH reaction in irradiated mice receiving splenocyte grafts at 20 mg/kg
Catalog No. 50-291-1302 Supplier Cayman Chemical Supplier No. 3480210 MG
An alkaloid; inhibits AChE activity (IC50 = 36.51 µM and scavenges ABTS radicals in cell-free assays from 12.5-100 µM; induces apoptosis in PANC-1 pancreatic cancer cells and reduces tumor growth in a PANC-1 mouse xenograft model; inhibits the GVH reaction in irradiated mice receiving splenocyte grafts at 20 mg/kg