Learn More
Medchemexpress LLC Phen green SK diacetate | 234075-45-7 | 95.3% | 706.48 g/mol | C37H21Cl2N3O8 | 5 MG
SDP
Catalog No. 5000264408
Encompass_Preferred
Phen Green SK diacetate is a cell-permeable, metal-ion-sensitive fluorescent probe used to detect and monitor intracellular chelatable metal ions. Chelation of Fe2+ and other transition metals causes fluorescence quenching; fluorescence can be reversed with membrane-permeable chelators, enabling assessment of intracellular metal dynamics. Reported excitation/emission maxima are approximately 507/532 nm.
  • Cell-permeable probe suitable for live-cell imaging.
  • Sensitive to transition and heavy metal ions such as Fe2+ and Zn2+.
  • Fluorescence quenching upon metal binding enables monitoring of metal levels.
  • Excitation/emission maxima near 507 nm/532 nm for fluorescence detection.
  • Reported purity around 95% as provided by supplier data.
  • Available in small milligram pack sizes for research applications.
Catalog No. 50-002-64408 Supplier Medchemexpress LLC Supplier No. HY1268235MG
Phen Green SK diacetate is a cell-permeable, metal-ion-sensitive fluorescent probe used to detect and monitor intracellular chelatable metal ions. Chelation of Fe2+ and other transition metals causes fluorescence quenching; fluorescence can be reversed with membrane-permeable chelators, enabling assessment of intracellular metal dynamics. Reported excitation/emission maxima are approximately 507/532 nm.
  • Cell-permeable probe suitable for live-cell imaging.
  • Sensitive to transition and heavy metal ions such as Fe2+ and Zn2+.
  • Fluorescence quenching upon metal binding enables monitoring of metal levels.
  • Excitation/emission maxima near 507 nm/532 nm for fluorescence detection.
  • Reported purity around 95% as provided by supplier data.
  • Available in small milligram pack sizes for research applications.