Learn More
Medchemexpress LLC Phen Green SK diacetate (5/6-mixture) | 234075-45-7 | 95.3% | 1 MG
SDP
Catalog No. 5000338904
Encompass_Preferred
Phen Green SK diacetate (5/6-mixture) is a metal ion-sensitive fluorescent probe that can penetrate cell membranes. It reacts with various metal ions like Fe2+, Cd2+, Co2+, Ni2+, and Zn2+. When Phen Green SK diacetate chelates Fe2+, it causes fluorescence quenching, which can be reversed by adding a membrane-permeable chelator, reflecting changes in the intracellular chelatable iron pool.
  • Metal ion-sensitive fluorescent probe.
  • Can penetrate cell membranes.
  • Reacts with a variety of metal ions including Fe2+, Cd2+, Co2+, Ni2+, and Zn2+.
  • Chelates Fe2+, leading to fluorescence quenching that can be restored, indicating changes in the intracellular chelatable iron pool.
  • Excitation/emission maxima are 507/532 nm.
  • Used to detect intracellular Fe2+ levels.
  • Can be used with a lipid peroxidation indicator dye to assess the ferroptosis state of cells.
Catalog No. 50-003-38904 Supplier Medchemexpress LLC Supplier No. HY1268231MG
Phen Green SK diacetate (5/6-mixture) is a metal ion-sensitive fluorescent probe that can penetrate cell membranes. It reacts with various metal ions like Fe2+, Cd2+, Co2+, Ni2+, and Zn2+. When Phen Green SK diacetate chelates Fe2+, it causes fluorescence quenching, which can be reversed by adding a membrane-permeable chelator, reflecting changes in the intracellular chelatable iron pool.
  • Metal ion-sensitive fluorescent probe.
  • Can penetrate cell membranes.
  • Reacts with a variety of metal ions including Fe2+, Cd2+, Co2+, Ni2+, and Zn2+.
  • Chelates Fe2+, leading to fluorescence quenching that can be restored, indicating changes in the intracellular chelatable iron pool.
  • Excitation/emission maxima are 507/532 nm.
  • Used to detect intracellular Fe2+ levels.
  • Can be used with a lipid peroxidation indicator dye to assess the ferroptosis state of cells.