Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phen-nadpo

Catalog No. 913758100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-375-8100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-375-8100MG Supplier Sigma Aldrich Supplier No. 913758100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phen-NaDPO is a universal cathode modifier for organic optoelectronics. Phen-NaDPO can significantly lower the work function of the Ag metal as well as ITO and HOPG.It lowers the energy loss between organic layer and the metal electrode, results in higher acheiveable efficiency for organic solar cells. It is also used as cathode interfacial material for inverted Perovskite solar cells, and electron transport materials for Perovskite solar cells and other organic electronic devices in general.

Specifications

CAS 1480371-38-7
Molecular Formula C34H23N2OP
Linear Formula C34H23N2OP
MDL Number MFCD31811570
Quantity 100 mg
Synonym 3-[6-(diphenylphosphinyl)-2-naphthalenyl]-1,10-Phenanthroline
Molecular Weight (g/mol) 506.53
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.