Learn More
Medchemexpress LLC Phenacetin-13C | 72156-72-0 | 99.9% | 100 MG
SDP
Catalog No. 5000406719
Encompass_Preferred
Phenacetin-13C is the 13C labeled Phenacetin. Phenacetin (Acetophenetidin) is a non-opioid analgesic/antipyretic agent and a selective COX-3 inhibitor. It is used as a probe of cytochrome P450 enzymes CYP1A2 in human liver microsomes and in rats.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development
Catalog No. 50-004-06719 Supplier Medchemexpress LLC Supplier No. HYB0476S1100MG
Phenacetin-13C is the 13C labeled Phenacetin. Phenacetin (Acetophenetidin) is a non-opioid analgesic/antipyretic agent and a selective COX-3 inhibitor. It is used as a probe of cytochrome P450 enzymes CYP1A2 in human liver microsomes and in rats.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes are incorporated into drug molecules for quantitation during drug development