Learn More
Medchemexpress LLC Phenacetin-13C | 72156-72-0 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0476S1
Encompass_Preferred
Phenacetin-13C is a 13C labeled version of Phenacetin, also known as Acetophenetidin. It is a non-opioid analgesic and antipyretic agent that acts as a selective COX-3 inhibitor. It is utilized as a probe for cytochrome P450 enzymes CYP1A2 in human liver microsomes and in rats. Stable heavy isotopes, such as carbon-13, are incorporated into drug molecules for quantitation during drug development, with deuteration also having potential impacts on pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Selective COX-3 inhibitor
  • Probe for cytochrome P450 enzymes CYP1A2
  • Features a stable heavy isotope (carbon-13) for quantitation during drug development
Catalog No. 50-004-50638