Learn More
Apexbio Technology LLC Phenacetin 62-44-2 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B14535.1
Encompass_Preferred
Phenacetin (CAS 62-44-2) is a synthetic non-opioid analgesic historically utilized for alleviating pain and reducing fever lacking intrinsic anti-inflammatory effects Its analgesic mechanism primarily involves inhibition of cyclooxygenase (COX) enzyme activity in the central nervous system leading to decreased prostaglandin synthesis Previously studied for its pharmacological action on pain modulation pathways phenacetin serves currently as an experimental reference compound in evaluating nephrotoxic effects in biomedical research following its market withdrawal due to nephropathy concerns
Catalog No. 50-000-63428