Learn More
Medchemexpress LLC HY-B0476 500mg Medchemexpress, Phenacetin CAS:62-44-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0476500MG
Encompass_Preferred
Medchemexpress, HY-B0476 500mg Phenacetin CAS:62-44-2 Phenacetin (Acetophenetidin) is a non-opioid analgesic/antipyretic agent. Phenacetin is a selective COX-3 inhibitor. Phenacetin is used as probe of cytochrome P450 enzymes CYP1A2 in human liver microsomes and in rats. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2552