Learn More
Medchemexpress LLC Phenacetin-d5 (Acetophenetidin-d5) | 69323-74-6 | 99.5% | 1 MG
SDP
Catalog No. 5000407096
Encompass_Preferred
Phenacetin-d5 is a deuterium-labeled form of Phenacetin, a non-opioid analgesic and antipyretic. It acts as a selective COX-3 inhibitor and is utilized as a probe for cytochrome P450 enzymes. This compound is a white to off-white solid with a molecular weight of 184.25 and a chemical formula of C10H8D5NO2.
  • Used as a probe of cytochrome P450 enzymes CYP1A2 in human liver microsomes and in rats.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-07096 Supplier Medchemexpress LLC Supplier No. HYB0476S1MG
Phenacetin-d5 is a deuterium-labeled form of Phenacetin, a non-opioid analgesic and antipyretic. It acts as a selective COX-3 inhibitor and is utilized as a probe for cytochrome P450 enzymes. This compound is a white to off-white solid with a molecular weight of 184.25 and a chemical formula of C10H8D5NO2.
  • Used as a probe of cytochrome P450 enzymes CYP1A2 in human liver microsomes and in rats.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.