Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenacetin-d5 (Acetophenetidin-d5) | 69323-74-6 | MFCD04118296 | 99.5% | 184.25 g/mol | C10H8D5NO2 | 10 MG
SDP

Catalog No. 5000297102
Encompass_Preferred

Phenacetin-d5 is a deuterium-labeled analogue of phenacetin used as an analytical standard and internal standard in mass spectrometry and quantitative assays. Supplied as a 10 mg solid with high chemical and isotopic purity, it is suitable for method development, quantitative analysis, and pharmacokinetic studies.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High analytical purity (>99.5%).
  • Provided as a 10 mg solid suitable for reconstitution.
  • Molecular weight 184.25 g/mol and formula C10H8D5NO2.
  • Useful for quantitative assays and method validation.

Catalog No. 50-002-97102 Supplier Medchemexpress LLC Supplier No. HYB0476S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenacetin-d5 is a deuterium-labeled analogue of phenacetin used as an analytical standard and internal standard in mass spectrometry and quantitative assays. Supplied as a 10 mg solid with high chemical and isotopic purity, it is suitable for method development, quantitative analysis, and pharmacokinetic studies.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High analytical purity (>99.5%).
  • Provided as a 10 mg solid suitable for reconstitution.
  • Molecular weight 184.25 g/mol and formula C10H8D5NO2.
  • Useful for quantitative assays and method validation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.