Learn More
U.S. Pharmacopeia Phenacetin Melting Point Standard, (Approximately 135 degrees), 62-44-2, MFCD00009094, 1g
Catalog No. NC9849083
Encompass
Molecular Formula C10H13NO2, Molecular Weight 179.218, Melting point/freezing point 273.02 - 280.22 °F (133.9 - 137.9 °C), Initial boiling point and boiling range 467.6 - 473 °F (242 - 245 °C), Synonyms: p-Acetophenetidide, Acetphenetidin, N-(4-Ethoxyphenyl) acetamide)
This item is not returnable. View return policy
Catalog No. NC9849083 Supplier U.S. Pharmacopeia Supplier No. 1514008
Molecular Formula C10H13NO2, Molecular Weight 179.218, Melting point/freezing point 273.02 - 280.22 °F (133.9 - 137.9 °C), Initial boiling point and boiling range 467.6 - 473 °F (242 - 245 °C), Synonyms: p-Acetophenetidide, Acetphenetidin, N-(4-Ethoxyphenyl) acetamide)