Learn More
Cayman Chemical Phenanthrene, Purity: ≥98%, CAS: 85-01-8, 100 g
SDP
Catalog No. 502949798
Encompass_Preferred
A polycyclic aromatic hydrocarbon; accumulates in the environment, is toxic to aquatic species, and is considered a pollutant; has been used as an intermediate in the synthesis of certain pesticides, plastics, and steroids
Catalog No. 50-294-9798 Supplier Cayman Chemical Supplier No. 34284100 G
A polycyclic aromatic hydrocarbon; accumulates in the environment, is toxic to aquatic species, and is considered a pollutant; has been used as an intermediate in the synthesis of certain pesticides, plastics, and steroids