Learn More
Medchemexpress LLC HY-B1727 100mg Medchemexpress, Phenanthrene CAS:85-01-8 Purity:>98%
SDP
Catalog No. 501970159
Encompass_Preferred
Medchemexpress, HY-B1727 100mg Phenanthrene CAS:85-01-8 Phenanthrene is a polycyclic aromatic hydrocarbon (PAH) and has been frequently used as an indicator for monitoring PAH contaminated matrices. Phenanthrene induces oxidative stress and inflammation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0159 Supplier Medchemexpress LLC Supplier No. HYB1727100MG
Medchemexpress, HY-B1727 100mg Phenanthrene CAS:85-01-8 Phenanthrene is a polycyclic aromatic hydrocarbon (PAH) and has been frequently used as an indicator for monitoring PAH contaminated matrices. Phenanthrene induces oxidative stress and inflammation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.