Learn More
Apexbio Technology LLC Phenanthrene 85-01-8 200mg
SDP
Catalog No. 5000090154
Encompass_Preferred
Phenanthrene is a polycyclic aromatic hydrocarbon (PAH) that exerts its biological activity primarily through the induction of cellular inflammatory responses enhancement of reactive oxygen species (ROS) generation and activation of apoptotic signaling pathways Based on these pharmacological properties phenanthrene is utilized in research to study PAH-induced cellular toxicity oxidative stress mechanisms and programmed cell death pathways Additionally it serves as a representative analyte in environmental toxicology investigations to detect quantify and evaluate PAH contamination in water and soil supporting the development of toxicological evaluation models and detection methods
Catalog No. 50-000-90154 Supplier Apexbio Technology LLC Supplier No. C6921200
Phenanthrene is a polycyclic aromatic hydrocarbon (PAH) that exerts its biological activity primarily through the induction of cellular inflammatory responses enhancement of reactive oxygen species (ROS) generation and activation of apoptotic signaling pathways Based on these pharmacological properties phenanthrene is utilized in research to study PAH-induced cellular toxicity oxidative stress mechanisms and programmed cell death pathways Additionally it serves as a representative analyte in environmental toxicology investigations to detect quantify and evaluate PAH contamination in water and soil supporting the development of toxicological evaluation models and detection methods