Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenanthrene-d10 | 1517-22-2 | MFCD00039418 | 99.5% | 188.30 g/mol | C14D10 | 10 MG
SDP

Catalog No. 5000296656
Encompass_Preferred

Phenanthrene-d10 is the deuterium-labeled analog of phenanthrene supplied for research use as an isotopically labeled internal standard and tracer for quantitative analysis by NMR, GC-MS, and LC-MS. It is intended for laboratory research and environmental analysis applications and is not for human or clinical use.

  • Deuterated internal standard for quantitative NMR, GC-MS, and LC-MS.
  • Molecular formula C14D10; molecular weight 188.30 g/mol.
  • High purity (≈99.5%) for accurate quantitation.
  • CAS number 1517-22-2 for unambiguous identification.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).

Catalog No. 50-002-96656 Supplier Medchemexpress LLC Supplier No. HYB1727S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenanthrene-d10 is the deuterium-labeled analog of phenanthrene supplied for research use as an isotopically labeled internal standard and tracer for quantitative analysis by NMR, GC-MS, and LC-MS. It is intended for laboratory research and environmental analysis applications and is not for human or clinical use.

  • Deuterated internal standard for quantitative NMR, GC-MS, and LC-MS.
  • Molecular formula C14D10; molecular weight 188.30 g/mol.
  • High purity (≈99.5%) for accurate quantitation.
  • CAS number 1517-22-2 for unambiguous identification.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.