Learn More
Medchemexpress LLC Phenanthrene-d10 | 1517-22-2 | MFCD00039418 | 99.5% | 25 MG
SDP
Catalog No. 5000414183
Encompass_Preferred
Phenanthrene-d10 is the deuterium-labeled Phenanthrene, a polycyclic aromatic hydrocarbon (PAH) frequently used as an indicator for monitoring PAH contaminated matrices. It induces oxidative stress and inflammation and is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-14183 Supplier Medchemexpress LLC Supplier No. 25MGHYB1727S
Phenanthrene-d10 is the deuterium-labeled Phenanthrene, a polycyclic aromatic hydrocarbon (PAH) frequently used as an indicator for monitoring PAH contaminated matrices. It induces oxidative stress and inflammation and is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS