Learn More
Medchemexpress LLC Phenanthrene-d10 | 1517-22-2 | MFCD00039418 | 99.5% | 5 MG
SDP
Catalog No. 5000413180
Encompass_Preferred
Phenanthrene-d10 is a deuterium labeled polycyclic aromatic hydrocarbon (PAH) frequently used as an indicator for monitoring PAH contaminated matrices. It induces oxidative stress and inflammation.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-13180 Supplier Medchemexpress LLC Supplier No. HYB1727S5MG
Phenanthrene-d10 is a deuterium labeled polycyclic aromatic hydrocarbon (PAH) frequently used as an indicator for monitoring PAH contaminated matrices. It induces oxidative stress and inflammation.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.