Learn More
Apexbio Technology LLC PHENANTHRENE-D10-50MG
SDP
Catalog No. 5000570236
Encompass_Preferred
Phenanthrene-d10 (CAS No 1517-22-2) is a deuterated polycyclic aromatic hydrocarbon used in biomedical research It serves as a stable isotopic label aiding in the evaluation of metabolic pathways and environmental studies Its mechanism often involves the cytochrome P450 enzymatic pathway enabling the study of xenobiotic metabolism In vitro activities are typically present in the nanomolar to low micromolar range making it suitable for precise metabolic analyses This compound is widely applied in cell models and animal studies to monitor various physiological processes and is a valuable tool in drug screening assays particularly when investigating the pharmacokinetics of candidate molecules Experimental concentrations typically depend on the specific research design ensuring flexibility across different experimental setups
Catalog No. 50-005-70236 Supplier Apexbio Technology LLC Supplier No. SL103150
Phenanthrene-d10 (CAS No 1517-22-2) is a deuterated polycyclic aromatic hydrocarbon used in biomedical research It serves as a stable isotopic label aiding in the evaluation of metabolic pathways and environmental studies Its mechanism often involves the cytochrome P450 enzymatic pathway enabling the study of xenobiotic metabolism In vitro activities are typically present in the nanomolar to low micromolar range making it suitable for precise metabolic analyses This compound is widely applied in cell models and animal studies to monitor various physiological processes and is a valuable tool in drug screening assays particularly when investigating the pharmacokinetics of candidate molecules Experimental concentrations typically depend on the specific research design ensuring flexibility across different experimental setups