Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenanthrene-d10, Quantitative, 1000 μg/mL, Methylene Chloride, 1 mL

Catalog No. USIST2301
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-IST-2301 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-IST-2301 Supplier Agilent Technologies Supplier No. IST2301
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1517-22-2 , 75-09-2
Molecular Formula C14H10
Molecular Weight (g/mol) 188.295
InChI Key YNPNZTXNASCQKK-LHNTUAQVSA-N
PubChem CID 519070
IUPAC Name 1,2,3,4,5,6,7,8,9,10-decadeuteriophenanthrene
SMILES C1=CC=C2C(=C1)C=CC3=CC=CC=C32

Specifications

CAS Max % 99.92, 0.08
Analyte or Component Names Dichloromethane,phenanthrene-d10
Element Dichloromethane, Phenanthrene-d10
Solvent or Matrix Dichloromethane
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Phenanthrene-d10 Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.