Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenanthrene, For environmental analysis, MilliporeSigma™ Supelco™

Catalog No. 111019100 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9100 5000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9100 Supplier MilliporeSigma Supelco Supplier No. 48569
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phenanthrene is a polycyclic aromatic hydrocarbon often formed by incomplete combustion of organic materials.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 85-01-8
Density 1.063 g/mL (at 25°C)
Physical Form Neat
Linear Formula C14H10
MDL Number MFCD00001168
Melting Point 98°C to 100°C
Boiling Point 340°C
Type Organic Pollutant Standard
Industry Type Environmental Testing
Quantity 5000 mg
Formula Weight 178.23
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.