Learn More
Medchemexpress LLC Phenanthrene (Standard) | 85-01-8 | 99.6% | 5 G
SDP
Catalog No. 5000412418
Encompass_Preferred
Phenanthrene (Standard) is the analytical standard of Phenanthrene, intended for research and analytical applications. It is an orally active polycyclic aromatic hydrocarbon (PAH) that induces inflammation, oxidative stress, and apoptosis. Additionally, phenanthrene is commonly used to detect or assess PAH pollution in the environment.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-004-12418 Supplier Medchemexpress LLC Supplier No. HYB17275G
Phenanthrene (Standard) is the analytical standard of Phenanthrene, intended for research and analytical applications. It is an orally active polycyclic aromatic hydrocarbon (PAH) that induces inflammation, oxidative stress, and apoptosis. Additionally, phenanthrene is commonly used to detect or assess PAH pollution in the environment.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.