Learn More
Medchemexpress LLC Phenanthrene (Standard) | 85-01-8 | 99.8% | 250 MG
SDP
Catalog No. 5000414492
Encompass_Preferred
Phenanthrene is an orally active polycyclic aromatic hydrocarbon (PAH) that induces inflammation, oxidative stress, and apoptosis. It is also commonly used to detect or assess PAH pollution in the environment.
  • Reduces mitochondrial membrane potential (MMP) in primary hepatocytes.
  • Induces oxidative stress and inflammation.
  • Causes liver damage and promotes hepatocyte apoptosis.
Catalog No. 50-004-14492 Supplier Medchemexpress LLC Supplier No. 250MGHYB1727R
Phenanthrene is an orally active polycyclic aromatic hydrocarbon (PAH) that induces inflammation, oxidative stress, and apoptosis. It is also commonly used to detect or assess PAH pollution in the environment.
  • Reduces mitochondrial membrane potential (MMP) in primary hepatocytes.
  • Induces oxidative stress and inflammation.
  • Causes liver damage and promotes hepatocyte apoptosis.