Learn More
Medchemexpress LLC Phenanthriplatin | 1416900-51-0 | 95.0% | 503.80 g/mol | C13H13ClN4O3Pt | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1398135MG
Encompass_Preferred
Phenanthriplatin is a monovalent platinum(II)-based cytotoxic complex used in research on DNA-targeting anticancer agents. It binds DNA and inhibits transcription, making it useful for cellular cytotoxicity assays and mechanistic studies of platinum-based compounds.
  • Monovalent platinum(II) complex that interacts with DNA.
  • Demonstrated potent cytotoxicity against cancer cell lines.
  • Useful for transcription inhibition and DNA-binding studies.
  • Supplied with stated purity suitable for research applications.
  • Available in small-mass packages for laboratory assays.
Catalog No. 50-002-22719