Learn More
Selleck Chemical LLC Phenanthroline monohydrate
SDP
Catalog No. 5000003109
Encompass_Preferred
1 10-Phenanthroline is a classic chelating bidentate ligand for transition metal ions that has played an important role in the development of coordination chemistry It is an inhibitor of metallopeptidases
Catalog No. 50-000-03109 Supplier Selleck Chemical LLC Supplier No. S5543100MG
1 10-Phenanthroline is a classic chelating bidentate ligand for transition metal ions that has played an important role in the development of coordination chemistry It is an inhibitor of metallopeptidases