Learn More
Medchemexpress LLC Phenazine-1-carboxylic acid | 2538-68-3 | 100.0% | 224.21 | 100 MG
SDP
Catalog No. 5000403993
Encompass_Preferred
Phenazine-1-carboxylic acid is an antifungal agent that also exhibits anticancer activity by inducing apoptosis in cancer cells through the regulation of ROS generation. It can upregulate the expression of IL-8 and ICAM-1 while inhibiting the release of RANTES and MCP-1, indicating potential immunomodulatory effects. It holds significant research value in anti-infection, anticancer, and immune response modulation.
  • Acts as an antifungal agent
  • Exhibits anticancer activity by inducing apoptosis
  • Regulates reactive oxygen species (ROS) generation
  • Upregulates IL-8 and ICAM-1 expression
  • Inhibits RANTES and MCP-1 release for potential immunomodulation
  • Inhibits proliferation of DU145 cells
  • Induces apoptosis through mitochondrial pathways
  • Prevents V. anguillarum infection in zebrafish embryos
Catalog No. 50-004-03993 Supplier Medchemexpress LLC Supplier No. HY33037100MG
Phenazine-1-carboxylic acid is an antifungal agent that also exhibits anticancer activity by inducing apoptosis in cancer cells through the regulation of ROS generation. It can upregulate the expression of IL-8 and ICAM-1 while inhibiting the release of RANTES and MCP-1, indicating potential immunomodulatory effects. It holds significant research value in anti-infection, anticancer, and immune response modulation.
  • Acts as an antifungal agent
  • Exhibits anticancer activity by inducing apoptosis
  • Regulates reactive oxygen species (ROS) generation
  • Upregulates IL-8 and ICAM-1 expression
  • Inhibits RANTES and MCP-1 release for potential immunomodulation
  • Inhibits proliferation of DU145 cells
  • Induces apoptosis through mitochondrial pathways
  • Prevents V. anguillarum infection in zebrafish embryos