Learn More
Medchemexpress LLC Phenazine-1-carboxylic acid | 2538-68-3 | 100.0% | 224.22 | 1 G
SDP
Catalog No. 5000403814
Encompass_Preferred
Phenazine-1-carboxylic acid is an antifungal agent that also exhibits anticancer activity by inducing apoptosis in cancer cells through the regulation of ROS generation. It can upregulate the expression of IL-8 and ICAM-1 while inhibiting the release of RANTES and MCP-1, demonstrating its potential immunomodulatory effects. It holds significant research value in anti-infection, anticancer, and immune response modulation.
  • Exhibits antineuroinflammatory activity by inhibiting NO production in LPS-induced mouse BV-2 cells
  • Inhibits proliferation of DU145 prostate cancer cells
  • Induces apoptosis in DU145 cells through ROS generation and mitochondrial apoptosis pathways
  • Upregulates IL-8 and ICAM-1 expression
  • Inhibits RANTES and MCP-1 release, showing immunomodulatory potential
  • Significantly prevents *Vibrio anguillarum* infection in zebrafish embryos
  • Increases embryo hatching rate in zebrafish models
Catalog No. 50-004-03814 Supplier Medchemexpress LLC Supplier No. HY330371G
Phenazine-1-carboxylic acid is an antifungal agent that also exhibits anticancer activity by inducing apoptosis in cancer cells through the regulation of ROS generation. It can upregulate the expression of IL-8 and ICAM-1 while inhibiting the release of RANTES and MCP-1, demonstrating its potential immunomodulatory effects. It holds significant research value in anti-infection, anticancer, and immune response modulation.
  • Exhibits antineuroinflammatory activity by inhibiting NO production in LPS-induced mouse BV-2 cells
  • Inhibits proliferation of DU145 prostate cancer cells
  • Induces apoptosis in DU145 cells through ROS generation and mitochondrial apoptosis pathways
  • Upregulates IL-8 and ICAM-1 expression
  • Inhibits RANTES and MCP-1 release, showing immunomodulatory potential
  • Significantly prevents *Vibrio anguillarum* infection in zebrafish embryos
  • Increases embryo hatching rate in zebrafish models