Learn More
Medchemexpress LLC Phenazine-1-carboxylic acid | 2538-68-3 | MFCD00078174 | 100.0% | 224.21 | 250 MG
SDP
Catalog No. 5000403103
Encompass_Preferred
Phenazine-1-carboxylic acid is an antifungal agent. It also shows anticancer activity by inducing apoptosis in cancer cells through the regulation of reactive oxygen species (ROS) generation. Additionally, it can upregulate the expression of IL-8 and ICAM-1 while inhibiting the release of RANTES and MCP-1, indicating potential immunomodulatory effects. This compound holds significant research value in anti-infection, anticancer, and immune response modulation studies.
  • Antifungal agent
  • Exhibits anticancer activity by inducing apoptosis through ROS generation
  • Upregulates IL-8 and ICAM-1 expression
  • Inhibits RANTES and MCP-1 release
  • Potential immunomodulatory effects
  • Research value in anti-infection, anticancer, and immune response modulation
  • Solid appearance
  • Light yellow to green yellow color
  • Initial source: Microorganisms, marine natural products, Truncatella angustata
Catalog No. 50-004-03103 Supplier Medchemexpress LLC Supplier No. 250MGHY33037
Phenazine-1-carboxylic acid is an antifungal agent. It also shows anticancer activity by inducing apoptosis in cancer cells through the regulation of reactive oxygen species (ROS) generation. Additionally, it can upregulate the expression of IL-8 and ICAM-1 while inhibiting the release of RANTES and MCP-1, indicating potential immunomodulatory effects. This compound holds significant research value in anti-infection, anticancer, and immune response modulation studies.
  • Antifungal agent
  • Exhibits anticancer activity by inducing apoptosis through ROS generation
  • Upregulates IL-8 and ICAM-1 expression
  • Inhibits RANTES and MCP-1 release
  • Potential immunomodulatory effects
  • Research value in anti-infection, anticancer, and immune response modulation
  • Solid appearance
  • Light yellow to green yellow color
  • Initial source: Microorganisms, marine natural products, Truncatella angustata