Learn More
Medchemexpress LLC Phenazine | 92-82-0 | MFCD00005023 | 100.0% | 180.21 g/mol | C12H8N2 | 100 G
SDP
Catalog No. 5000301140
Encompass_Preferred
Phenazine is an aromatic heterocyclic organic compound (C12H8N2, CAS 92-82-0, MW 180.21 g/mol) supplied as a high-purity analytical standard for research applications. It functions as a redox-active reagent and reference compound for biochemical assays and analytical chemistry.
  • Aromatic heterocyclic compound suitable for analytical and research use.
  • High purity confirmed by HPLC, appropriate for reference standard applications.
  • Molecular formula C12H8N2 and molecular weight 180.21 g/mol.
  • Functions as a redox reagent in biochemical and microbiological studies.
  • Stable solid form for routine storage and handling in laboratory settings.
  • Available in laboratory pack sizes for research and assay workflows.
Catalog No. 50-003-01140 Supplier Medchemexpress LLC Supplier No. HYW016477100G
Phenazine is an aromatic heterocyclic organic compound (C12H8N2, CAS 92-82-0, MW 180.21 g/mol) supplied as a high-purity analytical standard for research applications. It functions as a redox-active reagent and reference compound for biochemical assays and analytical chemistry.
  • Aromatic heterocyclic compound suitable for analytical and research use.
  • High purity confirmed by HPLC, appropriate for reference standard applications.
  • Molecular formula C12H8N2 and molecular weight 180.21 g/mol.
  • Functions as a redox reagent in biochemical and microbiological studies.
  • Stable solid form for routine storage and handling in laboratory settings.
  • Available in laboratory pack sizes for research and assay workflows.