Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phenazine | 92-82-0 | MFCD00005023 | 100.0% | 180.21 g/mol | C12H8N2 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYW016477

Encompass_Preferred

Phenazine is an aromatic heterocyclic compound (C12H8N2, MW 180.21 g/mol, CAS 92-82-0) supplied as a light yellow to yellow solid for research and analytical applications. It is used as a reference standard and a reagent in studies of redox-active natural products, dyes, and antimicrobial chemistry.

  • High purity (99.96%) suitable for analytical and research use.
  • Light yellow to yellow solid physical form for easy handling and storage.
  • Molecular formula C12H8N2 and molecular weight 180.21 g/mol for precise calculations.
  • Batch-specific certificate of analysis available for quality verification.
  • Recommended storage conditions provided to maintain stability.

Catalog No. 50-003-04201


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.