Learn More
Medchemexpress LLC HY-W004520 100mg Medchemexpress, Phenazine (methylsulfate) CAS:299-11-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW004520100MG
Encompass_Preferred
Medchemexpress, HY-W004520 100mg Phenazine (methylsulfate) CAS:299-11-6 Phenazine methylsulfate is a free radical generator. Phenazine methylsulfate has been used as an electron transfer reactant in cell viability assays. Phenazine methylsulfate induces ssDNA break formation in the presence of the reducing agent NADPH. Phenazine methylsulfate induces oxidative DNA damage in an alkaline comet assay and apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3222