Learn More
Medchemexpress LLC Phenazinium, 1-methoxy-5-methyl-, methyl sulfate (1:1) | 65162-13-2 | 99.3% | 336.36 | 200 MG
SDP
Catalog No. 5000366220
Encompass_Preferred
Methoxy-PMS (1-Methoxy PMS) is a stable electron-transport mediator and an active oxygen formation inducer. It facilitates electron transfer between NAD(P)H and tetrazolium dyes, demonstrating no cytotoxicity in cell culture media.
  • Acts as an active oxygen formation inducer
  • Functions as a stable electron-transport mediator
  • Transfers electrons from NAD(P)H to tetrazolium dyes
  • Exhibits no cytotoxicity in cell culture media
  • Receives electrons from NADH or NADPH at the membrane or inside the cell
  • Passes electrons to WST-8 around the outer cell membrane
Catalog No. 50-003-66220 Supplier Medchemexpress LLC Supplier No. 200MGHYD0937
Methoxy-PMS (1-Methoxy PMS) is a stable electron-transport mediator and an active oxygen formation inducer. It facilitates electron transfer between NAD(P)H and tetrazolium dyes, demonstrating no cytotoxicity in cell culture media.
  • Acts as an active oxygen formation inducer
  • Functions as a stable electron-transport mediator
  • Transfers electrons from NAD(P)H to tetrazolium dyes
  • Exhibits no cytotoxicity in cell culture media
  • Receives electrons from NADH or NADPH at the membrane or inside the cell
  • Passes electrons to WST-8 around the outer cell membrane