Learn More
Medchemexpress LLC Phenazopyridine hydrochloride | 136-40-3 | 99.6% | 1 ML
SDP
Catalog No. 5000408051
Encompass_Preferred
Phenazopyridine hydrochloride is a competitive SARM1 inhibitor (IC50 145 μM) and a TRPM8 antagonist. It has local anesthetic and analgesic effects, and can promote neuronal differentiation. This product is used to relieve painful symptoms of conditions like cystitis and urethritis, and in the study of traumatic brain injury, peripheral neuropathy, and neurodegenerative diseases.
  • Competitive SARM1 inhibitor (IC50 145 μM)
  • TRPM8 antagonist
  • Exhibits local anesthetic and analgesic effects
  • Relieves painful symptoms of cystitis and urethritis
  • Promotes neuronal differentiation
  • Used in research for traumatic brain injury, peripheral neuropathy, and neurodegenerative diseases
Catalog No. 50-004-08051 Supplier Medchemexpress LLC Supplier No. HYB098510MM/1ML
Phenazopyridine hydrochloride is a competitive SARM1 inhibitor (IC50 145 μM) and a TRPM8 antagonist. It has local anesthetic and analgesic effects, and can promote neuronal differentiation. This product is used to relieve painful symptoms of conditions like cystitis and urethritis, and in the study of traumatic brain injury, peripheral neuropathy, and neurodegenerative diseases.
  • Competitive SARM1 inhibitor (IC50 145 μM)
  • TRPM8 antagonist
  • Exhibits local anesthetic and analgesic effects
  • Relieves painful symptoms of cystitis and urethritis
  • Promotes neuronal differentiation
  • Used in research for traumatic brain injury, peripheral neuropathy, and neurodegenerative diseases