Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenazopyridine hydrochloride, 99%, analytical standard

Catalog No. AC441921000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC441921000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC441921000 Supplier Thermo Scientific Chemicals Supplier No. 441921000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 136-40-3 | C11H11N5·ClH | 249.7 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-40-3
Molecular Formula C11H11N5·ClH
Molecular Weight (g/mol) 249.7
InChI Key QQBPIHBUCMDKFG-UHFFFAOYSA-N
Synonym phenazopyridine hydrochloride, phenazopyridine hcl, urodine, pyridium, pyripyridium, bisteril, diridone, mallophene, phenazodine, pyrazodine
PubChem CID 8691
ChEBI CHEBI:71419
IUPAC Name 3-phenyldiazenylpyridine-2,6-diamine;hydrochloride
SMILES C1=CC=C(C=C1)N=NC2=C(N=C(C=C2)N)N.Cl

Specifications

Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5 to max. 101.5% min
Quantity 100 mg
Merck Index 15,7324
Formula Weight 249.7
Percent Purity 99%
Grade Analytical
Physical Form Liquid
Chemical Name or Material Phenazopyridine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer if inhaled.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If bre
EINECSNumber 205-243-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.