Learn More
Cayman Chemical Phenazopyridine (hydrochloride), Purity: ≥98%, CAS: 136-40-3, 50 g
SDP
Catalog No. 502990474
Encompass_Preferred
An azo dye with analgesic properties; decreases the bladder distension-induced firing rate of afferent bladder Aγ-fibers, but not C-fibers, in anesthetized rats at 0.3, 1, and 3 mg/kg
Catalog No. 50-299-0474 Supplier Cayman Chemical Supplier No. 2968350 G
An azo dye with analgesic properties; decreases the bladder distension-induced firing rate of afferent bladder Aγ-fibers, but not C-fibers, in anesthetized rats at 0.3, 1, and 3 mg/kg