Learn More
Cayman Chemical Phenazopyridine (hydrochloride), Purity: ≥98%, CAS: 136-40-3, 5 g
SDP
Catalog No. 502988935
Encompass_Preferred
An azo dye with analgesic properties; decreases the bladder distension-induced firing rate of afferent bladder Aγ-fibers, but not C-fibers, in anesthetized rats at 0.3, 1, and 3 mg/kg
Catalog No. 50-298-8935 Supplier Cayman Chemical Supplier No. 296835 G
An azo dye with analgesic properties; decreases the bladder distension-induced firing rate of afferent bladder Aγ-fibers, but not C-fibers, in anesthetized rats at 0.3, 1, and 3 mg/kg