Learn More
Abbott Phencyclidine (PCP) Urine Dip Test
Catalog No. 22911146
Change view
Click to view available options
Detectable Analytes:
Phencyclidine (PCP)
Catalog No. Detectable Analytes
22-911-146 Phencyclidine (PCP)
1 options
This item is not returnable. View return policy
Catalog No. 22-911-146 Supplier Abbott Supplier No. DPC114
Add to Cart
Add to Cart
This item is not returnable. View return policy
A rapid, one step urine dip test for qualitative detection of PCP in human urine
  • Detects PCP at or above a cutoff of 25 ng/mL
  • Results in 5 min.
  • Built in test control
  • Results stable for up to 60 min.
Specifications
Detectable Analytes Phencyclidine (PCP)
Sample Type Human Urine
Test Time 5 min.