Learn More
Medchemexpress LLC Phenelzine-d5 sulfate | 1219798-40-9 | 99.97% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB1018AS1MG
Encompass_Preferred
Phenelzine-d5 sulfate is the deuterium labeled version of Phenelzine sulfate, an orally active, irreversible antidepressant agent. It inhibits monoamine oxidase (MAO-A and MAO-B), GABA transaminase, primary amine oxidase (PrAO), and sequesters reactive aldehydes. It also inhibits LSD1 (Ki: 5.6 μM), reduces oxidative stress, lipogenesis, and increases neurotransmitter levels. It is studied for potential in neurological, metabolic, and cancer-related diseases.
  • Used as a tracer (deuterium labeled compound).
  • Internal standard for quantitative analysis (NMR, GC-MS, LC-MS).
  • High purity (99.97%).
  • For research use only.
  • Potential neuroprotective effects.
  • Inhibits multiple enzymes (MAO-A, MAO-B, GABA transaminase, PrAO, LSD1).
  • Elevates neurotransmitter levels.
Catalog No. 50-004-07462