Learn More
Apexbio Technology LLC Phenelzine sulfate 156-51-4 500mg
SDP
Catalog No. 5000093830
Encompass_Preferred
Phenelzine sulfate (CAS 156-51-4) is a small-molecule inhibitor of monoamine oxidase (MAO) targeting both MAO-A and MAO-B isoforms By blocking MAO activity phenelzine sulfate limits the enzymatic degradation of monoamine neurotransmitters including norepinephrine serotonin and dopamine resulting in elevated synaptic levels This modulation of neurotransmitter availability underpins its utility in neuropharmacological research on mood regulation neurochemical signaling and mechanistic studies of neurotransmitter dynamics Phenelzine sulfate is commonly utilized to investigate the molecular and cellular effects of altered monoaminergic transmission in neuroscience and psychiatric research models
Catalog No. 50-000-93830 Supplier Apexbio Technology LLC Supplier No. A9913500
Phenelzine sulfate (CAS 156-51-4) is a small-molecule inhibitor of monoamine oxidase (MAO) targeting both MAO-A and MAO-B isoforms By blocking MAO activity phenelzine sulfate limits the enzymatic degradation of monoamine neurotransmitters including norepinephrine serotonin and dopamine resulting in elevated synaptic levels This modulation of neurotransmitter availability underpins its utility in neuropharmacological research on mood regulation neurochemical signaling and mechanistic studies of neurotransmitter dynamics Phenelzine sulfate is commonly utilized to investigate the molecular and cellular effects of altered monoaminergic transmission in neuroscience and psychiatric research models