Learn More
Medchemexpress LLC Phenelzine sulfate | 156-51-4 | 99.96% | 500 MG
SDP
Catalog No. 5000407527
Encompass_Preferred
This antidepressant agent is an irreversible, orally active inhibitor of monoamine oxidase (MAO-A and MAO-B). It also inhibits GABA transaminase, primary amine oxidase (PrAO), and LSD1, while sequestering reactive aldehydes. The compound elevates neurotransmitter levels and suppresses oxidative stress and lipogenesis. It is under investigation for various neurological, metabolic, and cancer-related conditions.
  • Irreversible, orally active inhibitor of MAO-A and MAO-B.
  • Inhibits GABA transaminase and primary amine oxidase (PrAO).
  • Sequester reactive aldehydes.
  • Inhibits LSD1 (Ki: 5.6 μM).
  • Suppresses oxidative stress and lipogenesis.
  • Elevates neurotransmitters (serotonin, norepinephrine, dopamine).
  • Investigated for neurological, metabolic, and cancer diseases (e.g., depression, anxiety, stroke, Parkinson's, Alzheimer's).
  • Purity: 99.96%.
Catalog No. 50-004-07527 Supplier Medchemexpress LLC Supplier No. HYB1018A500MG
This antidepressant agent is an irreversible, orally active inhibitor of monoamine oxidase (MAO-A and MAO-B). It also inhibits GABA transaminase, primary amine oxidase (PrAO), and LSD1, while sequestering reactive aldehydes. The compound elevates neurotransmitter levels and suppresses oxidative stress and lipogenesis. It is under investigation for various neurological, metabolic, and cancer-related conditions.
  • Irreversible, orally active inhibitor of MAO-A and MAO-B.
  • Inhibits GABA transaminase and primary amine oxidase (PrAO).
  • Sequester reactive aldehydes.
  • Inhibits LSD1 (Ki: 5.6 μM).
  • Suppresses oxidative stress and lipogenesis.
  • Elevates neurotransmitters (serotonin, norepinephrine, dopamine).
  • Investigated for neurological, metabolic, and cancer diseases (e.g., depression, anxiety, stroke, Parkinson's, Alzheimer's).
  • Purity: 99.96%.