Learn More
Selleck Chemical LLC Phenelzine sulfate-E0165-100MG
SDP
Catalog No. 503136795
Encompass_Preferred
Phenelzine Sulfate is a hydrazine derivative and acts as a potent non-selective monoamine oxidase (MAO) inhibitor with an IC50 of 0 9 M
Catalog No. 50-313-6795 Supplier Selleck Chemical LLC Supplier No. E0165100MG
Phenelzine Sulfate is a hydrazine derivative and acts as a potent non-selective monoamine oxidase (MAO) inhibitor with an IC50 of 0 9 M