Learn More
Selleck Chemical LLC Phenelzine sulfate-E0165-1G
SDP
Catalog No. 503138288
Encompass_Preferred
Phenelzine Sulfate is a hydrazine derivative and acts as a potent non-selective monoamine oxidase (MAO) inhibitor with an IC50 of 0 9 M
Catalog No. 50-313-8288 Supplier Selleck Chemical LLC Supplier No. E01651G
Phenelzine Sulfate is a hydrazine derivative and acts as a potent non-selective monoamine oxidase (MAO) inhibitor with an IC50 of 0 9 M