Learn More
TARGETMOL CHEMICALS INC Phenethyl ferulate 5MG
SDP
Catalog No. 502879531
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Phenethyl ferulate (Caffeic Acid 3-Methyl Phenethyl Ester) has chemopreventive properties against chemically induced colon carcinogenesis. It enhances apoptosis in azoxymethane induced colon tumors. It shows inhibitory activity against cyclooxygenase (COX) and 5-lipoxygenase (5-LOX) with IC50 values of 4.35 uM and 5.75 uM, respectively. Purity 99.58%
Catalog No. 50-287-9531 Supplier TARGETMOL CHEMICALS INC Supplier No. TN23305MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Phenethyl ferulate (Caffeic Acid 3-Methyl Phenethyl Ester) has chemopreventive properties against chemically induced colon carcinogenesis. It enhances apoptosis in azoxymethane induced colon tumors. It shows inhibitory activity against cyclooxygenase (COX) and 5-lipoxygenase (5-LOX) with IC50 values of 4.35 uM and 5.75 uM, respectively. Purity 99.58%