Learn More
Medchemexpress LLC Phenethyl ferulate | 71835-85-3 | 99.66% | 298.33 | 10 MG
SDP
Catalog No. 5000452628
Encompass_Preferred
Phenethyl ferulate is a major constituent of Qianghuo, and exhibits inhibitory activity against cyclooxygenase (COX) and 5-lipoxygenase (5-LOX). It is suitable for various laboratory applications requiring precise inhibition of these enzymes.
  • Inhibits cyclooxygenase (COX)
  • Inhibits 5-lipoxygenase (5-LOX)
  • White to off-white solid appearance
  • High purity for reliable results
Catalog No. 50-004-52628 Supplier Medchemexpress LLC Supplier No. HYW00924810MG
Phenethyl ferulate is a major constituent of Qianghuo, and exhibits inhibitory activity against cyclooxygenase (COX) and 5-lipoxygenase (5-LOX). It is suitable for various laboratory applications requiring precise inhibition of these enzymes.
  • Inhibits cyclooxygenase (COX)
  • Inhibits 5-lipoxygenase (5-LOX)
  • White to off-white solid appearance
  • High purity for reliable results