Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenethyl isothiocyanate, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019732 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9732 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9732 Supplier MilliporeSigma Supelco Supplier No. 68488250MG

missing translation for 'validMainframeMsg'

Specifications

CAS 2257-09-2
Density 1.094 g/mL (at 25°C)
Flash Point 113°C - closed cup
Linear Formula C6H5 CH2 CH2 NCS
MDL Number MFCD00004821
Boiling Point 139°C to 140°C (11 mmHg)
Synonym 2-Phenylethyl isothiocyanate
Type Analytical Standard
Quantity 250 mg
Formula Weight 163.24
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.