Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phenethylzinc Bromide Solution

Catalog No. 49930750ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-930-750ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 49-930-750ML Supplier Sigma Aldrich Supplier No. 49930750ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phenethylzinc bromide can be used as a reagent: In the transition metal-catalyzed Negishi cross-coupling reaction to prepare aryl or heteroaryl derivatives by reacting with organic halides or triflates via carbon-carbon bond formation. To prepare (4-phenyl-1-butyn-1-yl)benzene by reacting with phenylacetylene via palladium-catalyzed cross-coupling reaction. To synthesize benzyl substituted thienopyrimidine derivatives.Product of Rieke Metals, Inc. Rieke is a registered trademark of Rieke Metals, Inc.

Specifications

CAS 26681-88-9
Density 0.979 g/mL at 25°C
Boiling Point 55°C (0.05 mmHg, lit.)
Molecular Formula C10H11P
Linear Formula C6H5P(CH=CH2)2
Quantity 50 mL
MDL Number MFCD00048142
Molecular Weight (g/mol) 162.17 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.