Learn More
Apexbio Technology LLC Phenformin HCl, 834-28-6, MFCD00035039, 10 g
SDP
Supplier:  Apexbio Technology LLC B137310000
Encompass_Preferred
MF: C10H16ClN5, MW: 241.72, Solubility: 12.1mg/mL in DMSO. Phenformin HCl is a hydrochloride salt of the biguanidine drug phenformin. Phenformin is the biguanide class that displays anti-diabetic activity. In 5 patients with diabetes mellitus, phenformin reduced the blood glucose values and glycosuria. In 4 of 5 patients, cholesterol and total lipid levels were dropped. Also, Phenformin had no impact on glucose, nor the insulin levels after intravenous glucose loading.
Catalog No. 50-101-0326