Learn More
Selleck Chemical LLC Phenformin HCl 50mg 834-28-6
SDP
Catalog No. 505792
Encompass_Preferred
Phenformin HCl is a hydrochloride salt of phenformin that is an anti-diabetic drug from the biguanide class. It activates AMPK, increasing activity and phosphorylation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-579-2 Supplier Selleck Chemical LLC Supplier No. S254250MG
Phenformin HCl is a hydrochloride salt of phenformin that is an anti-diabetic drug from the biguanide class. It activates AMPK, increasing activity and phosphorylation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.